Learn More
Sigma Aldrich Fine Chemicals Biosciences Accutase solution, MFCD01632905, 100mL
Catalog No. NC1670906
Encompass
Accutase solution (in dulbecco′s phosphate-buffered saline (DPBS) without Ca++ or Mg++) contains proteolytic and collagenolytic enzymes for cell detachment. It is useful for the routine detachment of cells from standard tissue culture plasticware and adhesion coated plasticware.
Catalog No. NC1670906 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A6964100ML
Accutase solution (in dulbecco′s phosphate-buffered saline (DPBS) without Ca++ or Mg++) contains proteolytic and collagenolytic enzymes for cell detachment. It is useful for the routine detachment of cells from standard tissue culture plasticware and adhesion coated plasticware.