Learn More
Innovative Cell Technologies AccutaseGMP 500ML, GMP-Grade Cell Detachment Solution, Non-Mammalian Enzyme Mixture
SDP
Catalog No. NC3800889
Encompass
AccutaseGMP is a GMP-grade cell detachment solution from Innovative Cell Technologies, Inc. It provides a gentle and efficient method for dissociating adherent cells, ensuring high viability without the need for washing or inactivation steps. Ideal for sensitive cell types, its formulation simplifies cell passaging, while its rigorous quality standards guarantee safety and purity. Available in a ready-to-use 500 ml volume, it supports both research and manufacturing needs.
Catalog No. NC3800889 Supplier Innovative Cell Technologies Supplier No. AT107500ML
AccutaseGMP is a GMP-grade cell detachment solution from Innovative Cell Technologies, Inc. It provides a gentle and efficient method for dissociating adherent cells, ensuring high viability without the need for washing or inactivation steps. Ideal for sensitive cell types, its formulation simplifies cell passaging, while its rigorous quality standards guarantee safety and purity. Available in a ready-to-use 500 ml volume, it supports both research and manufacturing needs.