Learn More
ACCUTEC BLADES INC (3P) AccuTec PRO Steel Back Single Edge Razor Blade, 100 Pack SKU# APBL-7064-0000 100 Blades Individually Wrapped in a Clamshell, 50 Clamshells per Master Case
Supplier:  ACCUTEC BLADES INC (3P) APBL70640000
Encompass_Preferred
Scrape, trim, or cut, it's all easier with AccuTec PRO Single Edge Razor Blades .009" high carbon steel and steel backed for added durability. Ideal for removing paint and decals from windows, cleaning a glass stovetop, or industrial uses such as removing material buildup on machinery. Single edge razor blade fits most standard single edge scrapers and box cutters. Units per Case: 50
Catalog No. NC3721132