Learn More
Beaver Visitec Intl Ltd ACCU-TEMP CAUTERY, VARIABLE LOW TEMP, 0.5 FINE TIP, 10/BX
Catalog No. NC0100006
Encompass
Sterile, single-use. 10 per box. Accu-Temp Variable Low Temperature Cautery 454°-649°C (849°-1200°F), 1/2 in. shaft with fine tip
Catalog No. NC0100006 Supplier Beaver Visitec Intl Ltd Supplier No. 8440000
Sterile, single-use. 10 per box. Accu-Temp Variable Low Temperature Cautery 454°-649°C (849°-1200°F), 1/2 in. shaft with fine tip