Learn More
JANT PHARMACAL CORPORATION ACCUTEST-10 SplitCup (COC/AMP/mAMP/THC/MTD/OPI/PCP/BAR/BZO/MDMA) 25 Tests/Kit
SDP
Catalog No. NC3457068
Encompass_Preferred
The ACCUTEST Urine Drug SplitCup combines the drug screens to provide a rapid, one step screening test for the simultaneous, qualitative detection of multiple drugs and drug metabolites in human urine. The splitcup is a split specimen drug testing solution that provides sample integrity.
Catalog No. NC3457068 Supplier JANT PHARMACAL CORPORATION Supplier No. DS07AC625
The ACCUTEST Urine Drug SplitCup combines the drug screens to provide a rapid, one step screening test for the simultaneous, qualitative detection of multiple drugs and drug metabolites in human urine. The splitcup is a split specimen drug testing solution that provides sample integrity.