Learn More
JANT PHARMACAL CORPORATION ACCUTEST Urine Reagent Strips 4-parameter test for Ketone, Protein, Glucose and pH. 100 test bottle.
SDP
Catalog No. NC3457024
Encompass_Preferred
Accutest URS-4 Urine Reagent Strips for Urinalysis are in vitro diagnostic test devices that use reagents for qualitative and semi-quantitative urinalysis. Accutest URS-4 Urine Reagent Strips are for single use in professional near patient (point-of-care) facilities and centralized laboratory locations by medical technologists read visually.Accutest URS-4 Urine Reagent Strips for Urinalysis are intended for use to detect conditions indicating possible metabolic abnormalities, liver diseases, kidney function, and urinary tract infections. Test results can be used along with other diagnostic information to rule out certain disease states and to determine if microscopic analysis is needed.
Catalog No. NC3457024 Supplier JANT PHARMACAL CORPORATION Supplier No. UA774
Accutest URS-4 Urine Reagent Strips for Urinalysis are in vitro diagnostic test devices that use reagents for qualitative and semi-quantitative urinalysis. Accutest URS-4 Urine Reagent Strips are for single use in professional near patient (point-of-care) facilities and centralized laboratory locations by medical technologists read visually.Accutest URS-4 Urine Reagent Strips for Urinalysis are intended for use to detect conditions indicating possible metabolic abnormalities, liver diseases, kidney function, and urinary tract infections. Test results can be used along with other diagnostic information to rule out certain disease states and to determine if microscopic analysis is needed.