Learn More
JANT PHARMACAL CORPORATION ACCUTEST MultiDrug Screen- 5 panel(Amphetamine, Cocaine, Methamphetamine, THC, Opiates (at 2,000ng/mL) 25 tests/kit
SDP
Catalog No. NC3457051
Encompass_Preferred
The ACCUTEST multi drug urine screen dip test card is a one-step urine drug screen that provides a rapid, one step screening test for the simultaneous, qualitative detection of multiple drugs and drug metabolites in human urine.
Catalog No. NC3457051 Supplier JANT PHARMACAL CORPORATION Supplier No. DS01AC425
The ACCUTEST multi drug urine screen dip test card is a one-step urine drug screen that provides a rapid, one step screening test for the simultaneous, qualitative detection of multiple drugs and drug metabolites in human urine.