Learn More
JANT PHARMACAL CORPORATION ACCUTEST MultiDrug Screen-5 w. COC, M-AMP, MOP(300), BZO, THC 25 Tests/Kit
SDP
Catalog No. NC3457052
Encompass_Preferred
The ACCUTEST multi drug urine screen dip test card is a one-step urine drug screen that provides a rapid, one step screening test for the simultaneous, qualitative detection of multiple drugs and drug metabolites in human urine.
Catalog No. NC3457052 Supplier JANT PHARMACAL CORPORATION Supplier No. DS134AC425
The ACCUTEST multi drug urine screen dip test card is a one-step urine drug screen that provides a rapid, one step screening test for the simultaneous, qualitative detection of multiple drugs and drug metabolites in human urine.