Learn More
JANT PHARMACAL CORPORATION ACCUTEST-5 Split Cup (COC/ mAMP/ THC/ OPI/ AMP) *CLIA Waived 25 Tests/Kit
SDP
Catalog No. NC3457064
Encompass_Preferred
The ACCUTEST Urine Drug SplitCup combines the drug screens to provide a rapid, one step screening test for the simultaneous, qualitative detection of multiple drugs and drug metabolites in human urine. The splitcup is a split specimen drug testing solution that provides sample integrity.
Catalog No. NC3457064 Supplier JANT PHARMACAL CORPORATION Supplier No. DS02AC625
The ACCUTEST Urine Drug SplitCup combines the drug screens to provide a rapid, one step screening test for the simultaneous, qualitative detection of multiple drugs and drug metabolites in human urine. The splitcup is a split specimen drug testing solution that provides sample integrity.