Learn More
JANT PHARMACAL CORPORATION ACCUTEST SplitCup-6+3 w. Adulteration Screen (pH/ S.G./ Oxidant) (AMP1000 /COC 300 /MET 1000/ OPI 2000/ PCP 25/ THC 50) 25 Tests/Kit
SDP
Catalog No. NC3457065
Encompass_Preferred
The ACCUTEST Urine Drug SplitCup combines the drug screens to provide a rapid, one step screening test for the simultaneous, qualitative detection of multiple drugs and drug metabolites in human urine. The splitcup is a split specimen drug testing solution that provides sample integrity.
Catalog No. NC3457065 Supplier JANT PHARMACAL CORPORATION Supplier No. DS08AC625
The ACCUTEST Urine Drug SplitCup combines the drug screens to provide a rapid, one step screening test for the simultaneous, qualitative detection of multiple drugs and drug metabolites in human urine. The splitcup is a split specimen drug testing solution that provides sample integrity.