Learn More
JANT PHARMACAL CORPORATION ACCUTEST SplitCup-8+6 w. Adulteration Screen (pH/ S.G./ Oxidant/Nit/Glut/Creatinine)(AMP/BAR/BZO/COC/MAMP/OPI/PCP/THC) 25 Tests/Kit
SDP
Catalog No. NC3457076
Encompass_Preferred
The ACCUTEST Urine Drug SplitCup combines the drug screens to provide a rapid, one step screening test for the simultaneous, qualitative detection of multiple drugs and drug metabolites in human urine. The splitcup is a split specimen drug testing solution that provides sample integrity.
Catalog No. NC3457076 Supplier JANT PHARMACAL CORPORATION Supplier No. DS208
The ACCUTEST Urine Drug SplitCup combines the drug screens to provide a rapid, one step screening test for the simultaneous, qualitative detection of multiple drugs and drug metabolites in human urine. The splitcup is a split specimen drug testing solution that provides sample integrity.