Learn More
ACCUTEC BLADES INC (3P) AccuThrive Double Edge Pathology Blade SKU# AVBL-3000-0000 Carbon Blade, 100 Blades per Carton, 10 Cartons per Case
Catalog No. NC3721106
Encompass_Preferred
The AccuThrive Double Edge Pathology Blade is designed to help pathologists hand trim specimens. Designed for use prior to microtome use, our disposable pathology blades are intended for use in gross dissection of tissue samples. Units per Case: 10
Catalog No. NC3721106 Supplier ACCUTEC BLADES INC (3P) Supplier No. AVBL30000000
The AccuThrive Double Edge Pathology Blade is designed to help pathologists hand trim specimens. Designed for use prior to microtome use, our disposable pathology blades are intended for use in gross dissection of tissue samples. Units per Case: 10