Learn More
Seracare Life Sciences Inc AccuTrak™ HBV DNA Genotype Qualification Panel, 9 Members, Research Use Only, 9 x 1.2 mL
Catalog No. NC3652979
Encompass
The AccuTrak™ HBV DNA Genotype Qualification Panel is a 9-member validation tool essential for developing and evaluating hepatitis B virus genotype test methods. This panel includes eight HBV DNA positive plasma samples and one negative sample, providing dependable material for optimal assay performance. Its Research Use Only status reinforces its significance for diagnostic manufacturers and clinical laboratories in advancing viral diagnostics.
Catalog No. NC3652979 Supplier Seracare Life Sciences Inc Supplier No. 24000225
The AccuTrak™ HBV DNA Genotype Qualification Panel is a 9-member validation tool essential for developing and evaluating hepatitis B virus genotype test methods. This panel includes eight HBV DNA positive plasma samples and one negative sample, providing dependable material for optimal assay performance. Its Research Use Only status reinforces its significance for diagnostic manufacturers and clinical laboratories in advancing viral diagnostics.