Learn More
JANT PHARMACAL CORPORATION Accutest H. pylori Urease Test, Qualitative Rapid Urease Test, 50 tests/box, Room Temperature Storage, ClIA Waived
SDP
Catalog No. NC3348353
Encompass_Preferred
The Accutest H. pylori Urease Test is a qualitative rapid urease test designed for the quick detection of *Helicobacter pylori* from gastric mucosal biopsy samples in symptomatic adult patients. This CLIA-waived test provides a user-friendly solution with fast results, making it ideal for clinical decisions in endoscopy settings.
  • Provides positive results in as little as 1 minute.
  • Negative results confirmed within 1 hour.
  • No refrigeration needed; can be stored at room temperature.
  • High sensitivity (>99%) and specificity (>99%) for reliable diagnosis.
  • Simple procedure with no incubation or external reagents required.
  • Cost-effective with low cost per test.
  • Long shelf life of 3 years.
  • Includes controls to ensure test accuracy.
  • 50 tests per box.
Catalog No. NC3348353 Supplier JANT PHARMACAL CORPORATION Supplier No. ID750
The Accutest H. pylori Urease Test is a qualitative rapid urease test designed for the quick detection of *Helicobacter pylori* from gastric mucosal biopsy samples in symptomatic adult patients. This CLIA-waived test provides a user-friendly solution with fast results, making it ideal for clinical decisions in endoscopy settings.
  • Provides positive results in as little as 1 minute.
  • Negative results confirmed within 1 hour.
  • No refrigeration needed; can be stored at room temperature.
  • High sensitivity (>99%) and specificity (>99%) for reliable diagnosis.
  • Simple procedure with no incubation or external reagents required.
  • Cost-effective with low cost per test.
  • Long shelf life of 3 years.
  • Includes controls to ensure test accuracy.
  • 50 tests per box.