Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accuTupH™ Rugged Bulb pH Combination Electrodes PROMO

Catalog No. 1362018AS24 Shop All Fisherbrand Products
Change view
Click to view available options
Length (English):
4 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
13-620-18AS24 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-620-18AS24

missing translation for 'validMainframeMsg'

Rugged Bulb pH Combination Electrodes are mercury-free and optimal for lab use and precise Q.C. measurements with a response time of under 10 seconds.

  • pH combination electrode
  • Double junction
  • Temperature range: 80°C to 100°C
  • pH range: 0 to 14
TRUSTED_SUSTAINABILITY

Specifications

Connector Types BNC
Junction Double
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Combination pH Electrode
Filling Solution Type Saturated KCl
For Use With (Application) pH, Tris Buffer
Internal Reference Ag/AgCl
mV Range 0 to 25 mV
Response Time <10 sec.
Temperature (English) Operating 32°F to 212°F
Length (English) 4 in.
Length (Metric) 102 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.