Learn More
Beckman Coulter Accuvette™ ST Sampling Vials with Caps, 25 mL
Catalog No. 15655125
Change view
Click to view available options
Capacity (Metric):
25 mL
Catalog No. Capacity (Metric)
15-655-125 25 mL
1 options
Catalog No. 15-655-125 Supplier Beckman Coulter Supplier No. A35473/DEL
Designed for use on the stage of the Multisizer 4e to ensure optimal fluid use and circulation with the locked platform height.
  • Designed to ensure optimal fluid use and circulation with the locked platform height of the Multisizer 4e
  • Rounded corners in the interior encourage re-suspension of settled particles
  • Disposable
  • 200 Vials/Box
TRUSTED_SUSTAINABILITY
Specifications
Product Type Sampling Vial
Capacity (Metric) 25 mL
Material Polystyrene
For Use With (Equipment) Multisizer™ 4e Coulter Counters
Includes 200 Vials with Caps