Learn More
Med Vet International ACD-A 20ml in a Collapsible 50ml Bag, J1350A, Each
Catalog No. 5000581285
Encompass_Preferred
The rapidly increasing use of regenerative medicine modalities makes the demands for new ancillary products needed to complete these procedures.  The use of anticoagulants such as ACD-A are an important component for these new regenerative procedures such as PRP - Platelet Rich Plasma, Bone Marrow Aspirates, and Autologous Protein Solution. -20ml ACD-A • Sodium Citrate Dehydrate 2H2O • Citric Acid Monohydrate • Dextrose Monohydrate -50ml Fluid Bag • Single drug delivery port • Latex-free rubber port cap • Sterile   Package of 6 Bags   Weight and dimensions in additional information are for shipping purposes only.
Catalog No. 50-005-81285 Supplier Med Vet International Supplier No. JOR1350A
The rapidly increasing use of regenerative medicine modalities makes the demands for new ancillary products needed to complete these procedures.  The use of anticoagulants such as ACD-A are an important component for these new regenerative procedures such as PRP - Platelet Rich Plasma, Bone Marrow Aspirates, and Autologous Protein Solution. -20ml ACD-A • Sodium Citrate Dehydrate 2H2O • Citric Acid Monohydrate • Dextrose Monohydrate -50ml Fluid Bag • Single drug delivery port • Latex-free rubber port cap • Sterile   Package of 6 Bags   Weight and dimensions in additional information are for shipping purposes only.