Learn More
Aviva Systems Biology Corp ACDC Antibody - N-terminal region (ARP32799_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32799_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ACDC. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ACDC. Alternate names: ACDC, ADPN, APM1, APM-1, GBP28, ACRP30, ADIPQTL1ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-52875