Learn More
Cayman Chemical AcDEVDCHO trifluoroacetate s
SDP
Supplier:  Cayman Chemical 100175 MG
Encompass_Preferred
A potent, reversible inhibitor of caspase-3 and caspase-7 (Kis = 0.23 and 1.6 nM, respectively); also inhibits other caspases (Kis = 18, 1710, 132, 205, 31, 0.92, 60, and 12 nM for caspases-1, -2, -4, -5, -6, -8, -9, and -10, respectively)
Catalog No. 50-288-9337