Learn More
Cayman Chemical Ac-DEVD-CHO (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502887717
Encompass_Preferred
A potent, reversible inhibitor of caspase-3 and caspase-7 (Kis = 0.23 and 1.6 nM, respectively); also inhibits other caspases (Kis = 18, 1710, 132, 205, 31, 0.92, 60, and 12 nM for caspases-1, -2, -4, -5, -6, -8, -9, and -10, respectively)
Catalog No. 50-288-7717 Supplier Cayman Chemical Supplier No. 1001710 MG
A potent, reversible inhibitor of caspase-3 and caspase-7 (Kis = 0.23 and 1.6 nM, respectively); also inhibits other caspases (Kis = 18, 1710, 132, 205, 31, 0.92, 60, and 12 nM for caspases-1, -2, -4, -5, -6, -8, -9, and -10, respectively)