Learn More
Cayman Chemical Ac-DMQD-CHO (trifluoroacetate salt), Purity: ≥95%, 5 mg
SDP
Catalog No. 502939635
Encompass_Preferred
An inhibitor of caspase-3 (IC50 = 39 nM); selective for caspase-3 over caspase-1 and caspase-8 (IC50s = 148 and 1,100 nM, respectively); inhibits camptothecin-induced apoptosis in immortalized human T/C 28a4 chondrocytes and primary human chondrocytes; reduces the number of apoptotic cells, cavitation, and demyelination in a rat model of traumatic injury to the spinal cord at 1 mg/kg
Catalog No. 50-293-9635 Supplier Cayman Chemical Supplier No. 271035 MG
An inhibitor of caspase-3 (IC50 = 39 nM); selective for caspase-3 over caspase-1 and caspase-8 (IC50s = 148 and 1,100 nM, respectively); inhibits camptothecin-induced apoptosis in immortalized human T/C 28a4 chondrocytes and primary human chondrocytes; reduces the number of apoptotic cells, cavitation, and demyelination in a rat model of traumatic injury to the spinal cord at 1 mg/kg