Learn More
Apexbio Technology LLC ACDPP hydrochloride 1177808-75-1 10mg
SDP
Catalog No. 5000091231
Encompass_Preferred
ACDPP hydrochloride (CAS 1177808-75-1) is a selective antagonist of the metabotropic glutamate receptor 5 (mGluR5) By inhibiting mGluR5 signaling ACDPP hydrochloride modulates glutamatergic neurotransmission and has been utilized to investigate the molecular mechanisms underlying neurological and psychiatric disorders such as anxiety depression neuropathic pain and certain neurodegenerative diseases This compound serves as a valuable tool in preclinical studies focused on developing therapeutic strategies targeting mGluR5-mediated pathologies
Catalog No. 50-000-91231 Supplier Apexbio Technology LLC Supplier No. B699110
ACDPP hydrochloride (CAS 1177808-75-1) is a selective antagonist of the metabotropic glutamate receptor 5 (mGluR5) By inhibiting mGluR5 signaling ACDPP hydrochloride modulates glutamatergic neurotransmission and has been utilized to investigate the molecular mechanisms underlying neurological and psychiatric disorders such as anxiety depression neuropathic pain and certain neurodegenerative diseases This compound serves as a valuable tool in preclinical studies focused on developing therapeutic strategies targeting mGluR5-mediated pathologies