Learn More
Sigma Aldrich Fine Chemicals Biosciences Ace addition funnel capaci1EA
Catalog No. 502905066
Encompass
Angled position makes manipulation of stopcock easier than conventional style. Leak-proof with complete drainage pressure-equalizing arm precise needle-valve adjustment and double-scale graduations.Threaded stopcock (0 to 3 mm orifice for funnels up to 250 mL and 0 to 5 mm orifice for funnels 500 mL and larger) with PTfE plug permits smooth needle-valve adjustment down to 0.1 mL/min flow rate. Double PTfE-ring seals prevent exposure of backup O-ring to corrosive liquids.
Catalog No. 50-290-5066 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2567731EA
Angled position makes manipulation of stopcock easier than conventional style. Leak-proof with complete drainage pressure-equalizing arm precise needle-valve adjustment and double-scale graduations.Threaded stopcock (0 to 3 mm orifice for funnels up to 250 mL and 0 to 5 mm orifice for funnels 500 mL and larger) with PTfE plug permits smooth needle-valve adjustment down to 0.1 mL/min flow rate. Double PTfE-ring seals prevent exposure of backup O-ring to corrosive liquids.