Learn More
Ace Glass, Inc. ACE BURLITCH SYSTEM C ACE BURLITCH SYSTEM C
SDP
Catalog No. 50979475
Encompass
ACE-Burlitch inert atmosphere system, designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with Ace no-air labware, or suitiably adapted conventional glassware. Complete instructions on assembly, leak testing, and preparation of the system are included.
Catalog No. 50-979-475 Supplier Ace Glass, Inc. Supplier No. 781810
Add to Cart
Add to Cart
Request a Quote
ACE-Burlitch inert atmosphere system, designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with Ace no-air labware, or suitiably adapted conventional glassware. Complete instructions on assembly, leak testing, and preparation of the system are included.