Learn More
Sigma Aldrich Fine Chemicals Biosciences Ace giant Soxhlet extracti1EA
Catalog No. 502909117
Encompass
Complete with bulb type condenser and one flask. Cycling rate may be doubled over conventional extractors. All joints are interchangeable. Size C is supplied with adapter Z218871 for connecting the XL extractor joint and condenser. Order giant thimbles separately.
Catalog No. 50-290-9117 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2187581EA
Complete with bulb type condenser and one flask. Cycling rate may be doubled over conventional extractors. All joints are interchangeable. Size C is supplied with adapter Z218871 for connecting the XL extractor joint and condenser. Order giant thimbles separately.