Learn More
Ace Glass, Inc. Ace Glass Fully Jacketed Funnel, 1000 mL, #80 Ace-Thred Bottom
SDP
Catalog No. NC3990274
Encompass
This jacketed filtration apparatus is designed for use with 75 mm membranes or filter paper, making it ideal for applications requiring temperature control, such as solvent recycling. Its fully threaded design eliminates the need for traditional clamping.
  • Jacketed for temperature control.
  • Designed for 75 mm membranes or filter paper.
  • All threaded apparatus for clamp-free operation.
  • Coarse glass fritted disc, interchangeable with an all PTFE support.
  • Features a GL80 wide mouth bottle.
  • Includes a #15 Ace-Thred vacuum connection.
Catalog No. NC3990274 Supplier Ace Glass, Inc. Supplier No. 371011
This jacketed filtration apparatus is designed for use with 75 mm membranes or filter paper, making it ideal for applications requiring temperature control, such as solvent recycling. Its fully threaded design eliminates the need for traditional clamping.
  • Jacketed for temperature control.
  • Designed for 75 mm membranes or filter paper.
  • All threaded apparatus for clamp-free operation.
  • Coarse glass fritted disc, interchangeable with an all PTFE support.
  • Features a GL80 wide mouth bottle.
  • Includes a #15 Ace-Thred vacuum connection.