Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse ACE ELISA Kit

Catalog No. EMACE
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EMACE 96 Tests
EMACEX5 5 x 96 Tests
EMACEX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EMACE Supplier Invitrogen™ Supplier No. EMACE
Add to Cart
Add to Cart

ELISA

The Mouse Angiotensin Converting Enzyme (Ms ACE) ELISA quantitates Ms ACE in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms ACE. Principle of the method The Mouse ACE solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

This gene encodes an enzyme involved in catalyzing the conversion of angiotensin I into a physiologically active peptide angiotensin II. Angiotensin II is a potent vasopressor and aldosterone-stimulating peptide that controls blood pressure and fluid-electrolyte balance. This enzyme plays a key role in the renin-angiotensin system. Many studies have associated the presence or absence of a 287 bp Alu repeat element in this gene with the levels of circulating enzyme or cardiovascular pathophysiologies. Two most abundant alternatively spliced variants of this gene encode two isozymes-the somatic form and the testicular form that are equally active. Multiple additional alternatively spliced variants have been identified but their full length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09470
Assay Range 0.123-30 ng/mL
Assay Sensitivity 0.12 ng/mL
Calibration Range 0.12 ng/ml to 30 ng/ml
Content And Storage Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.
Assay Solid-phase sandwich ELISA
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Proteases Peptidases & Inhibitors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN-gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF.
Sufficient For 96 assays
Target Species Mouse
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias ACE; ACE1; ACE-T; angiotensin 1 converting enzyme 1; angiotensin converting enzyme; angiotensin converting enzyme, somatic isoform; angiotensin I converting enzyme; angiotensin I converting enzyme (peptidyl-dipeptidase A) 1; angiotensin I converting enzyme 1; angiotensin I converting enzyme peptidyl-dipeptidase A 1 transcript; angiotensin I-converting enzyme (Dipeptidyl carboxypeptidase 1); angiotensin-converting enzyme; Angiotensin-converting enzyme, soluble form; angiotensin-converting enzyme; LOW QUALITY PROTEIN: angiotensin-converting enzyme; angiotensin-converting enzyme-like; AW208573; carboxycathepsin; CD143; CD143 antigen; DCP; Dcp1; dipeptidyl carboxypeptidase 1; Dipeptidyl carboxypeptidase 1 (Angiotensin I-converting enzyme); dipeptidyl carboxypeptidase I; dipeptidyl peptidase; I79_009670; ICH; kininase II; LOC103158533; MGC26566; MVCD3; peptidase P; peptidyl dipeptidase i; StsRR92; testicular ECAACE; ACE1; ACE-T; angiotensin 1 converting enzyme 1; angiotensin converting enzyme; angiotensin converting enzyme, somatic isoform; angiotensin I converting enzyme; angiotensin I converting enzyme (peptidyl-dipeptidase A) 1; angiotensin I converting enzyme 1; angiotensin I converting enzyme peptidyl-dipeptidase A 1 transcript; angiotensin I-converting enzyme (Dipeptidyl carboxypeptidase 1); angiotensin-converting enzyme; Angiotensin-converting enzyme, soluble form; angiotensin-converting enzyme; LOW QUALITY PROTEIN: angiotensin-converting enzyme; angiotensin-converting enzyme-like; AW208573; carboxycathepsin; CD143; CD143 antigen; DCP; Dcp1; dipeptidyl carboxypeptidase 1; Dipeptidyl carboxypeptidase 1 (Angiotensin I-converting enzyme); dipeptidyl carboxypeptidase I; dipeptidyl peptidase; I79_009670; ICH; kininase II; LOC103158533; MGC26566; MVCD3; peptidase P; peptidyl dipeptidase i; StsRR92; testicular ECA; AW208573; CD143
Gene ID (Entrez) 11421
Gene Symbol Ace
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases ACE, Dipeptidyl carboxypeptidase I, Kininase II
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Cardiovascular Research
Sample Volume Plasma 1 μL, Serum 1 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym RP23-100A9.1, AW208573, CD143
Target ACE
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.