Learn More
Sigma Aldrich Fine Chemicals Biosciences Ace powder dispensing funn1EA
Catalog No. 502905253
Encompass
for powders and solids up to 25 mesh. features a flexible PTfE screw thread wrapped on a precision PTfE-ceramic shaft contained within a precision bore housing. Completely inert materials allow flushing with solvents. No.15 Ace-Thred affords easy disassembly and cleaning.Note This funnel operates on a screw feed principal. The rate or amount of powder added is controlled by turning the knob which feeds the powder into the funnel like a conveyor.
Catalog No. 50-290-5253 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2432481EA
for powders and solids up to 25 mesh. features a flexible PTfE screw thread wrapped on a precision PTfE-ceramic shaft contained within a precision bore housing. Completely inert materials allow flushing with solvents. No.15 Ace-Thred affords easy disassembly and cleaning.Note This funnel operates on a screw feed principal. The rate or amount of powder added is controlled by turning the knob which feeds the powder into the funnel like a conveyor.