Learn More
Ace Glass, Inc. Tube, pressure, 140mL, 150psi, 38.1mm OD, 20.3cm long, complete with #15 front-seal plug
SDP
Catalog No. NC9912854
Encompass_Preferred
120mL glass pressure tube with #15 Ace-Thred, rated at 150psig at as much as 120C. Includes #15 front-seal plug. Dimensions: 38.1mm O.D. x 20.3cm long.
Catalog No. NC9912854 Supplier Ace Glass, Inc. Supplier No. 864896
120mL glass pressure tube with #15 Ace-Thred, rated at 150psig at as much as 120C. Includes #15 front-seal plug. Dimensions: 38.1mm O.D. x 20.3cm long.