Learn More
Sigma Aldrich Fine Chemicals Biosciences Ace pressure tube bushing1EA
Catalog No. 502900385
Encompass
Heavy-wall borosilicate glass tubes are pressure rated to 150 psig. Useful for the polymerization and analysis of halogens by thermal decomposition in the presence of lime. Tubes are supplied complete with Ace-Thred PTfE bushing and fETfE(R) O-ring.for pressure work the front seal bushing is recommended (allows O-ring to sit at narrow neck of tube below the threads). Back seal bushing can also be used if preferred (allows O-ring to sit on top of the tube against cap).Replacement fETfE O-ringsfront SealZ504661 (Ace-Thred 7)Z504858 (Ace-Thred 15)Z505056 (Ace-Thred 25)Back SealZ504742 (Ace-Thred 7)Z505021 (Ace-Thred 15)Z505110 (Ace-Thred 25)CAUTION This glassware is not guaranteed against breakage caused by pressure or vacuum. These tubes should never be used if scratched or otherwise damaged.
Catalog No. 50-290-0385 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1810991EA
Heavy-wall borosilicate glass tubes are pressure rated to 150 psig. Useful for the polymerization and analysis of halogens by thermal decomposition in the presence of lime. Tubes are supplied complete with Ace-Thred PTfE bushing and fETfE(R) O-ring.for pressure work the front seal bushing is recommended (allows O-ring to sit at narrow neck of tube below the threads). Back seal bushing can also be used if preferred (allows O-ring to sit on top of the tube against cap).Replacement fETfE O-ringsfront SealZ504661 (Ace-Thred 7)Z504858 (Ace-Thred 15)Z505056 (Ace-Thred 25)Back SealZ504742 (Ace-Thred 7)Z505021 (Ace-Thred 15)Z505110 (Ace-Thred 25)CAUTION This glassware is not guaranteed against breakage caused by pressure or vacuum. These tubes should never be used if scratched or otherwise damaged.