Learn More
MARIAN MEDICAL INC ACE Stopper, 10Fr/30mm, W/ ACElok Dressing, 1 Each
Catalog No. NC3958251
Encompass_Preferred
ACE Stopper. Stoma Accessory. Marian Medical Inc. AP Series. Silicone ACE stopper for stoma closure and tract maintenance. Includes ACElok external fixation dressing. Available in multiple French sizes and lengths. For use in ACE and Malone stoma management. Packaged as 1 each.
Catalog No. NC3958251 Supplier MARIAN MEDICAL INC Supplier No. AP10/30
ACE Stopper. Stoma Accessory. Marian Medical Inc. AP Series. Silicone ACE stopper for stoma closure and tract maintenance. Includes ACElok external fixation dressing. Available in multiple French sizes and lengths. For use in ACE and Malone stoma management. Packaged as 1 each.