Learn More
MARIAN MEDICAL INC ACE Stopper, 14Fr/100mm, W/ ACElok Dressing, 1 Each
Catalog No. NC3958258
Encompass_Preferred
ACE Stopper. Stoma Accessory. Marian Medical Inc. AP Series. Silicone ACE stopper for stoma closure and tract maintenance. Includes ACElok external fixation dressing. Available in multiple French sizes and lengths. For use in ACE and Malone stoma management. Packaged as 1 each.
Catalog No. NC3958258 Supplier MARIAN MEDICAL INC Supplier No. AP14/100
ACE Stopper. Stoma Accessory. Marian Medical Inc. AP Series. Silicone ACE stopper for stoma closure and tract maintenance. Includes ACElok external fixation dressing. Available in multiple French sizes and lengths. For use in ACE and Malone stoma management. Packaged as 1 each.