Learn More
Mac-Mod Avantor ACE UltraCore C18-Amide, 95 A, 3.5 um, 4.6 x 150 mm Solid Core Column
SDP
Catalog No. 502367533
Encompass
ACE UltraCore 3.5 micron HPLC Columns are a unique particle size with core-shell architecture designed to provide higher efficiency than standard 3 micron fully porous material. ACE UltraCore 3.5 micron solid-core columns are available in a C18, Phenylhexyl, Biphenyl and C18-Amide bonding chemistry and a wide range of analytical column sizes. These ACE UltraCore columns are rated to 600 bar. Note Holder to fit ACE UltraCore column is BIOGRD0001.
Catalog No. 50-236-7533 Supplier Mac-Mod Supplier No. CORE35E1546
ACE UltraCore 3.5 micron HPLC Columns are a unique particle size with core-shell architecture designed to provide higher efficiency than standard 3 micron fully porous material. ACE UltraCore 3.5 micron solid-core columns are available in a C18, Phenylhexyl, Biphenyl and C18-Amide bonding chemistry and a wide range of analytical column sizes. These ACE UltraCore columns are rated to 600 bar. Note Holder to fit ACE UltraCore column is BIOGRD0001.