Learn More
Sigma Aldrich Fine Chemicals Biosciences Ace vapor duct tube for co1EA
Catalog No. 502900364
Encompass
The vapor duct tube accessories are for BAchi condenser V assemblies and will fit either BAchi Rotavapor model R-210/R-215 or R-114/R-144.Combi-clips not included.
Catalog No. 50-290-0364 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6824461EA
The vapor duct tube accessories are for BAchi condenser V assemblies and will fit either BAchi Rotavapor model R-210/R-215 or R-114/R-144.Combi-clips not included.