Learn More
Enzo Life Sciences ACE1 (human), (recombinant) (10µg)
SDP
Catalog No. 502013945
Encompass_Preferred
Alternative name: Angiotensin-converting enzyme 1, CD143. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. Contains sodium chloride, ZnCl2, TRIS and glycerol. Source: Produced in NSO cells. Long Term Storage: -80°C.
Catalog No. 50-201-3945 Supplier Enzo Life Sciences Supplier No. ENZPRT1560010
Alternative name: Angiotensin-converting enzyme 1, CD143. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. Contains sodium chloride, ZnCl2, TRIS and glycerol. Source: Produced in NSO cells. Long Term Storage: -80°C.