Learn More
Sino Biological Anti-ACE2 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10108MM36F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ACE2 (rh ACE2; Catalog#10108-H08H; NP_068576.1; Met1-Ser740) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-228-2712