Learn More
Sino Biological ACE2 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10108T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ACE2 (rh ACE2; Catalog#10108-H31H; NP_068576.1; Met1-Ser740). ACE2 specific IgG was purified by Human ACE2 affinity chromatography.
Catalog No. 50-224-7567