Learn More
Cayman Chemical ACE2 asy Buffer10X 1ea
SDP
Catalog No. 502896835
Encompass_Preferred
ACE2 Assay Buffer (10X) has been tested and formulated to work exclusively with Cayman’s Cayman’s ACE2 Inhibitor Screening Assay Kit (Item No. 502100).
Catalog No. 50-289-6835 Supplier Cayman Chemical Supplier No. 5021011 EA
ACE2 Assay Buffer (10X) has been tested and formulated to work exclusively with Cayman’s Cayman’s ACE2 Inhibitor Screening Assay Kit (Item No. 502100).