Learn More
Epigentek Inc Recombinant ACE2 Protein with His and Avi Tag (100 µg)
SDP
Supplier:  Epigentek Inc E800081
Encompass_Preferred
Recombinant ACE2 Protein is produced by HEK293 cells expression system. The target protein is expressed with sequence (Gln18-Ser740) of human ACE2 (Accession #Q9BYF1) fused with a 6xHis, Avi tag at the C-terminus.
Catalog No. 50-201-9355