Learn More
Cayman Chemical ACE2-HRP ConjUGate 100dtn
SDP
Catalog No. 502894955
Encompass_Preferred
ACE2-HRP Conjugate has been tested and formulated to work exclusively with Cayman’s SARS-CoV-2 (B.1.1.7, Alpha Variant) Neutralizing Antibody SimpleDetect ELISA Kit (Item No. 502180).
Catalog No. 50-289-4955 Supplier Cayman Chemical Supplier No. 502182100 DTN
ACE2-HRP Conjugate has been tested and formulated to work exclusively with Cayman’s SARS-CoV-2 (B.1.1.7, Alpha Variant) Neutralizing Antibody SimpleDetect ELISA Kit (Item No. 502180).