Learn More
AdipoGen ACE2 (human):Fc (human) (rec.) (100 µg)
SDP
Supplier:  AdipoGen CHIB232006C100
Encompass_Preferred
Recombinant protein. Extracellular domain of human ACE2 (aa 1-740) is fused to the N-terminus of the Fc region of human IgG1. Source: HEK 293 cells. Endotoxin: <0.001EU/µg purified protein (LAL test). Lyophilized. Contains PBS. ACE2 converts angiotensin I to angiotensin 1-9, a peptide of unknown function, and angiotensin II to angiotensin 1-7, a vasodilator. ACE2 is involved in the regulation of systemic blood pressure and has direct effects on cardiac functions. It is expressed predominantly in endothelial cells of the lung, gut, heart and kidney. ACE2 together with the protease TMPRSS2 acts as a functional receptor for SARS coronavirus as well as for the new highly pathogenic coronavirus, 2019-nCoV/SARS-CoV-2, which is cause for pneumonia COVID-19. It has been shown that human recombinant soluble ACE2 can significantly block early stages of SARS-CoV-2 infections.
Catalog No. 50-205-4693