Learn More
AdipoGen ACE2 (human) (rec.) (50 µg)
SDP
Supplier:  AdipoGen AG40B0192C050
Encompass_Preferred
Recombinant Protein. Extracellular domain of human ACE2 (aa 18-740) are fused at the C-terminus to a FLAG-tag. Source: HEK 293 cells. Endotoxin: <0.01EU/µg purified protein (LAL test). Lyophilized. Contains PBS. Angiotensin-converting enzyme2 (ACE2) is an ectoenzyme (carboxypeptidase) with an extracellular catalytic domain that predominantly localizes at the plasma membrane and is thereby able to hydrolyze circulating peptides. ACE2 converts angiotensin I to angiotensin 1-9, a peptide of unknown function, and angiotensin II to angiotensin 1-7, a vasodilator. ACE2 is involved in the regulation of systemic blood pressure and has direct effects on cardiac functions. It is expressed predominantly in endothelial cells of the lung, gut, heart and kidney. ACE2 together with the protease TMPRSS2 acts as a functional receptor for SARS coronavirus as well as for the new highly pathogenic coronavirus, 2019-nCoV/SARS-CoV-2, which is cause for pneumonia COVID-19.
Catalog No. 50-205-3295