Learn More
AdipoGen ACE2 human pAb
SDP
Supplier:  AdipoGen AG25A0042C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human ACE2. Detects a band of ~90kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. ACE2 has direct effects on cardiac functions. It is expressed predominantly in vascular endothelial cells of the heart and kidney. ACE2 converts angiotensin I to angiotensin 1-9. ACE2 was also identified as a functional receptor for SARS coronavirus.
Catalog No. 50-256-3269