Learn More
Epigentek Inc ACE2 Monoclonal Antibody (100ul)
SDP
Supplier:  Epigentek Inc A73665100
Encompass_Preferred
Unconjugated Monoclonal Antibody Raised in Rabbit. Immunogen: Recombinant fusion protein containing a sequence corresponding to amino acids 18-250 of human ACE2 (NP_068576.1).. Tested Applications: WB, IF. Reactivity: Human, Mouse, Rat. Alternative Names: ACE2, ACEH, angiotensin I converting enzyme 2, ACE2, angiotensin I converting enzyme 2, angiotensin-converting enzyme 2; ACE-related carboxypeptidase; angiotensin I converting enzyme (peptidyl-dipeptidase A) 2; angiotensin-converting enzyme homolog; metalloprotease MPROT15; peptidyl-dipeptidase A.
Catalog No. 50-201-9317