Learn More
Epigentek Inc ACE2 Monoclonal Antibody (100ul)
SDP
Supplier:  Epigentek Inc A73666100
Encompass_Preferred
Unconjugated Monoclonal Antibody Raised in Rabbit. Immunogen: A synthesized peptide derived from human TMPRSS2. Tested Applications: WB, IHC. Reactivity: Human, Mouse, Rat. Alternative Names: TMPRSS2, PP9284, PRSS10, transmembrane protease, serine 2, TMPRSS2, transmembrane protease, serine 2, transmembrane protease serine 2; epitheliasin; serine protease 10.
Catalog No. 50-201-9319