Learn More
Sino Biological Recombinant Anti-ACE2 Neutralizing Antibody
Supplier:  Sino Biological 10108MM37100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ACE2 (rh ACE2; Catalog #10108-H08H; Met1-Ser740; NP_068576.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7591