Learn More
Enzo Life Sciences ACE2 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS8220100
Encompass_Preferred
Isotype: IgG. Immunogen: Peptide corresponding to 17 amino acids near the center of human ACE2. The immunogen is located within amino acids 180 - 230 of ACE2. Species reactivity: Human, Mouse, Rat. Applications: ELISA, IF, IHC, WB. Purity: Affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-201-3346