Learn More
Sino Biological Recombinant Human ACE2 Protein (His Tag), Amine-Labeled Biotinylated, HPLC-verified
Supplier:  Sino Biological 10108H08HB100
Encompass_Preferred
A DNA sequence encoding the human ACE2 (NP_068576.1) (Met1-Ser740) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6252