Learn More
Sino Biological Human ACE2 / Angiotensin-Converting Enzyme 2 Protein (Fc Tag) 100ug
Supplier:  Sino Biological 10108H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ser 740) of human ACE2 precursor (NP_068576.1) was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-5401