Learn More
Sino Biological Recombinant Human ACE2 Protein (mFc Tag), HPLC-verified
Supplier:  Sino Biological 10108H05H100
Encompass_Preferred
A DNA sequence encoding the human ACE2 (NP_068576.1) (Met1-Ser740) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-211-5403